missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3,5-difluorophenol, 99%, Thermo Scientific Chemicals

Product Code. p-7028747
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15483417 250 mg 250mg
11386519 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15483417 Supplier Thermo Scientific Alfa Aesar Supplier No. H26153.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 130191-91-2 | Molecular Formula: C6H3BrF2O | Molecular Weight: 208.99 g/mol

4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-3,5-difluorophenol is used as intermediates of Liquid Crystals. Reactant in the synthesis of difluoroethyleneoxy group liquid crystal.

Solubility
Sparingly Soluble (8.2E-5 g/L) (25°C).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130191-91-2
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD08062385
InChI Key HHGOLZGZHXELSW-UHFFFAOYSA-N
PubChem CID 7172027
IUPAC Name 4-bromo-3,5-difluorophenol
SMILES C1=C(C=C(C(=C1F)Br)F)O

Specifications

Melting Point 72°C to 78°C
Quantity 250 mg
Sensitivity Hygroscopic
Synonym 2,6-difluoro-4-hydroxybromobenzene, phenol, 4-bromo-3,5-difluoro, 4-bromo-3,5-difluoro-phenol, pubchem14520, intermediates-zcf02006, acmc-20a41x, 3,5-difluoro-4-bromophenol, phenol,4-bromo-3,5-difluoro
Solubility Information Sparingly Soluble (8.2E-5g/L) (25°C).
Formula Weight 208.99
Percent Purity 99%
Chemical Name or Material 4-Bromo-3,5-difluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.