missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)benzenesulfonamide, Thermo Scientific™

Product Code. 10139639
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10139639 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10139639 Supplier Thermo Scientific Maybridge Supplier No. MAY00224DE
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 351003-62-8
Packaging Amber Glass Bottle
Molecular Formula C7H5BrF3NO2S
Quantity 5 g
Synonym 4-bromo-2-trifluoromethyl benzenesulfonamide, 4-bromo-2-trifluoromethyl benzenesulphonamide, 4-bromo-2-trifluoromethyl benzene sulfonamide, pubchem11731, 4-bromo-2-trifluoromethyl-benzenesulfonamide, 4-bromo-2-trifluoromethylbenzene sulfonamide, benzenesulfonamide,4-bromo-2-trifluoromethyl, benzenesulfonamide, 4-bromo-2-trifluoromethyl
InChI Key AHFGKYBAYSEBPR-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)S(=O)(=O)N
IUPAC Name 4-bromo-2-(trifluoromethyl)benzenesulfonamide
Molecular Weight (g/mol) 304.081
PubChem CID 2778392
Formula Weight 304.08
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzenesulfonamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.