missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)aniline, 98%

Product Code. 11400123
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400123 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400123 Supplier Thermo Scientific Alfa Aesar Supplier No. A12146.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 445-02-3 | C7H5BrF3N | 240.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-02-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
MDL Number MFCD00064393
InChI Key PHXGKHTWEOPCEW-UHFFFAOYSA-N
Synonym 2-amino-5-bromobenzotrifluoride, 4-bromo-2-trifluoromethyl aniline, benzenamine, 4-bromo-2-trifluoromethyl, 4-bromo-2-trifluoromethyl-phenylamine, 2-amino-5-bromo trifluoromethyl benzene, 2-amino-5-bromo benzotrifluoride, 4-bromo-2-trifluoroaniline, 2-trifluoromethyl-4-bromoaniline, 4-bromo-alpha,alpha,alpha-trifluoro-o-toluidine, 4-bromo-2-trifluoromethyl-aniline
PubChem CID 67960
IUPAC Name 4-bromo-2-(trifluoromethyl)aniline
SMILES NC1=CC=C(Br)C=C1C(F)(F)F

Specifications

Density 1.71
Boiling Point 84°C to 85°C (5 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.532
Quantity 25 g
UN Number UN2810
Beilstein 2211504
Formula Weight 240.03
Percent Purity 98%
Chemical Name or Material 4-Bromo-2- (trifluoromethyl)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.