missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-thiophenecarboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 207.05 |
Percent Purity | 95% |
Chemical Name or Material | 4-Bromo-2-thiophenecarboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'