missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitro-6-(trifluoromethyl)aniline, 98%, Thermo Scientific™

Catalog Number 15478518
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
157026-18-1
C7H4BrF3N2O2
285.02
MFCD09835318
JWIAFKYOENFPNQ-UHFFFAOYSA-N
4-bromo-2-nitro-6-trifluoromethyl aniline, 2-amino-5-bromo-3-nitrobenzotrifluoride, 4-bromo-2-nitro-6-trifluoromethyl-phenylamine, 4-bromo-2-nitro-6-trifluoromethyl benzenamine, 4-bromo-2-trifluoromethyl-6-nitrobenzenamine, acmc-209df8, ksc910q9b, benzenamine,4-bromo-2-nitro-6-trifluoromethyl, 4-bromo-2-nitro-6-trifluoromethyl-benzenamine
10850672
4-bromo-2-nitro-6-(trifluoromethyl)aniline
NC1=C(C=C(Br)C=C1C(F)(F)F)[N+]([O-])=O
This item is not returnable. View return policy
157026-18-1
C7H4BrF3N2O2
285.02
MFCD09835318
JWIAFKYOENFPNQ-UHFFFAOYSA-N
4-bromo-2-nitro-6-trifluoromethyl aniline, 2-amino-5-bromo-3-nitrobenzotrifluoride, 4-bromo-2-nitro-6-trifluoromethyl-phenylamine, 4-bromo-2-nitro-6-trifluoromethyl benzenamine, 4-bromo-2-trifluoromethyl-6-nitrobenzenamine, acmc-209df8, ksc910q9b, benzenamine,4-bromo-2-nitro-6-trifluoromethyl, 4-bromo-2-nitro-6-trifluoromethyl-benzenamine
10850672
4-bromo-2-nitro-6-(trifluoromethyl)aniline
NC1=C(C=C(Br)C=C1C(F)(F)F)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 285.02
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-nitro-6-(trifluoromethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'