Learn More
4-Bromo-2-(methylthio)pyrimidine, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 959236-97-6 |
---|---|
Molecular Formula | C5H5BrN2S |
Molecular Weight (g/mol) | 205.073 |
MDL Number | MFCD09878932 |
InChI Key | TZSQAGYDWPPKPE-UHFFFAOYSA-N |
Synonym | 4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine |
PubChem CID | 45787531 |
IUPAC Name | 4-bromo-2-methylsulfanylpyrimidine |
SMILES | CSC1=NC=CC(=N1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15491179
|
Thermo Scientific Alfa Aesar
H64124.MD |
250 mg |
N/A
|
N/A | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
959236-97-6 | |
205.073 | |
TZSQAGYDWPPKPE-UHFFFAOYSA-N | |
45787531 | |
CSC1=NC=CC(=N1)Br |
C5H5BrN2S | |
MFCD09878932 | |
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine | |
4-bromo-2-methylsulfanylpyrimidine |
Specifications
959236-97-6 | |
Brown to Yellow | |
MFCD09878932 | |
21913366 | |
TZSQAGYDWPPKPE-UHFFFAOYSA-N | |
4-bromo-2-methylsulfanylpyrimidine | |
45787531 | |
97% | |
4-Bromo-2-(methylthio)pyrimidine |
30°C to 40°C | |
C5H5BrN2S | |
UN1759 | |
4-bromo-2-methylthio pyrimidine, 4-bromo-2-methylsulfanyl pyrimidine, pyrimidine, 4-bromo-2-methylthio, 4-bromo-2-methylsulfanyl-pyrimidine, 2-methylthio-4-bromopyrimidine | |
CSC1=NC=CC(=N1)Br | |
205.073 | |
205.07 | |
Low Melting Solid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' : -30° to -10°C
Recommended Storage : Store at -20°C