missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methylbenzene-1-sulfonyl chloride, 97%, Maybridge

Product Code. 10446092
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10446092 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10446092 Supplier Thermo Scientific Maybridge Supplier No. RDP00246DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 139937-37-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C7H6BrClO2S
Linear Formula Br(CH3)C6H3SO2Cl
MDL Number MFCD00219927
Quantity 1 g
Synonym 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072
InChI Key KNBSFUSBZNMAKU-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl
IUPAC Name 4-bromo-2-methylbenzenesulfonyl chloride
Molecular Weight (g/mol) 269.537
PubChem CID 2781338
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methylbenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.