missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-methylbenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
£74.30 - £291.59
Chemical Identifiers
CAS | 139937-37-4 |
---|---|
Molecular Formula | C7H6BrClO2S |
Molecular Weight (g/mol) | 269.55 |
InChI Key | KNBSFUSBZNMAKU-UHFFFAOYSA-N |
Synonym | 4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 |
PubChem CID | 2781338 |
IUPAC Name | 4-bromo-2-methylbenzenesulfonyl chloride |
SMILES | CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
Description
Chemical Identifiers
139937-37-4 | |
269.55 | |
4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
4-bromo-2-methylbenzenesulfonyl chloride |
C7H6BrClO2S | |
KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
2781338 | |
CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl |
Specifications
139937-37-4 | |
164.0°C (15.0 mmHg) | |
C7H6BrClO2S | |
4-bromo-2-methylbenzene-1-sulfonyl chloride, 4-bromo-2-methylbenzenesulphonyl chloride, 4-bromo-2-methyl-benzenesulfonyl chloride, 2-methyl-4-bromobenzenesulfonylchloride, 5-bromo-2-chlorosulphonyl toluene, 2-methyl-4-bromobenzenesulfonyl chloride, benzenesulfonylchloride, 4-bromo-2-methyl, pubchem5066, buttpark 9857-56, intermediates-zcf02072 | |
CC1=C(C=CC(=C1)Br)S(=O)(=O)Cl | |
269.55 | |
269.55 | |
4-Bromo-2-methylbenzene-1-sulfonyl chloride |
62.0°C to 67.0°C | |
96% min. (GC) | |
Br(CH3)C6H3SO2Cl | |
KNBSFUSBZNMAKU-UHFFFAOYSA-N | |
4-bromo-2-methylbenzenesulfonyl chloride | |
2781338 | |
97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Bromo-2-methylbenzene-1-sulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only