missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methyl-6-nitroaniline, 97%

Product Code. 11436867
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436867 1 g 1g
11446867 5 g 5g
11456867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436867 Supplier Thermo Scientific Alfa Aesar Supplier No. A14319.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77811-44-0 | C7H7BrN2O2 | 231.049 g/mol

It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the design and synthesis of CK2 inhibitors and in the docking studies of novel telmisartan-glitazone hybrid analogs for the treatment of metabolic syndrome.

Solubility
Slightly soluble in water and soluble in hot methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77811-44-0
Molecular Formula C7H7BrN2O2
Molecular Weight (g/mol) 231.049
MDL Number MFCD00052919
InChI Key ZXFVKFUXKFPUQJ-UHFFFAOYSA-N
Synonym 4-bromo-2-methyl-6-nitro-phenylamine, 4-bromo-2-methyl-6-nitrobenzenamine, 2-amino-5-bromo-3-nitrotoluene, 4-bromo-6-nitro-o-toluidine, benzenamine, 4-bromo-2-methyl-6-nitro, 4-bromo-2-methyl-6-nitrophenylamine, 4-bromo-2-methyl-6-nitro-aniline, pubchem3813, acmc-1bg8u, ksc496e7t
PubChem CID 522679
IUPAC Name 4-bromo-2-methyl-6-nitroaniline
SMILES CC1=CC(=CC(=C1N)[N+](=O)[O-])Br

Specifications

Melting Point 143°C to 147°C
Quantity 1 g
Beilstein 3530524
Solubility Information Slightly soluble in water and soluble in hot methanol.
Formula Weight 231.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methyl-6-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.