missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxypyridine, 98%

Product Code. p-7055445
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441049 1 g 1g
15451049 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441049 Supplier Thermo Scientific Alfa Aesar Supplier No. H64026.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100367-39-3 | C6H6BrNO | 188.024 g/mol

4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-methoxypyridine used in the synhesis of Imidazo [1, 2-a] pyrimidines. Suzuki coupling between 4-methoxyphenylboronic acid and 4-bromo-2-methoxypyridine affords 1, 5-disubstituted pyridones. Starting material used in a synthesis of crown-ester-bipyridines and viologens via sodium or nickel reductive coupling, side chain oxidation and esterification.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100367-39-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD08277268
InChI Key YFTGMMXMLPTTAY-UHFFFAOYSA-N
Synonym 2-methoxy-4-bromopyridine, 4-bromo-2-methoxy-pyridine, pyridine, 4-bromo-2-methoxy, pubchem6657, ksc494g1d, acmc-1c790, abbypharma ap-17-5965
PubChem CID 14062309
IUPAC Name 4-bromo-2-methoxypyridine
SMILES COC1=NC=CC(=C1)Br

Specifications

Melting Point 25°C to 26°C
Color White to Yellow
Boiling Point 87°C (15 mmHg)
Quantity 1 g
Beilstein 115199
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 4-Bromo-2-methoxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.