missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxyphenol, 98%

Product Code. 11362716
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362716 2 g 2g
11372716 10 g 10g
11382716 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362716 Supplier Thermo Scientific Alfa Aesar Supplier No. L08801.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7368-78-7 | C7H7BrO2 | 203.035 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7368-78-7
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD00051937
InChI Key WHSIIJQOEGXWSN-UHFFFAOYSA-N
Synonym 4-bromoguaiacol, 5-bromo-2-hydroxyanisole, 4-bromo-2-methoxy-phenol, phenol, 4-bromo-2-methoxy, 2-methoxy-4-bromophenol, 5bromoguaiacol, 5-bromoguaiacol, 5-bromguajacol, pubchem17271, acmc-209orr
PubChem CID 262234
IUPAC Name 4-bromo-2-methoxyphenol
SMILES COC1=C(C=CC(=C1)Br)O

Specifications

Melting Point 32°C to 37°C
Boiling Point 129°C to 132°C (12 mmHg)
Odor Phenol-like
Quantity 2 g
Beilstein 2045286
Solubility Information Slightly soluble in water.
Formula Weight 203.04
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.