missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-hydroxypyridine, 97%

Product Code. 15454747
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15454747 5 g 5g
15464747 250 mg 250mg
15444747 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15454747 Supplier Thermo Scientific Alfa Aesar Supplier No. H32067.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36953-37-4 | C5H4BrNO | 173.997 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36953-37-4
Molecular Formula C5H4BrNO
Molecular Weight (g/mol) 173.997
MDL Number MFCD00234041
InChI Key SSLMGOKTIUIZLY-UHFFFAOYSA-N
Synonym 4-bromo-2-hydroxypyridine, 4-bromopyridin-2-ol, 4-bromopyridin-2 1h-one, 4-bromo-2-pyridinol, 4-bromo-1,2-dihydropyridin-2-one, 4-bromo-2 1h-pyridinone, 4-bromo-2-pyridone, 4-bromo-2-hydroxy pyridine, 2-hydroxy-4-bromopyridine, 2 1h-pyridinone, 4-bromo
PubChem CID 817115
IUPAC Name 4-bromo-1H-pyridin-2-one
SMILES C1=CNC(=O)C=C1Br

Specifications

Melting Point 201°C to 204°C
Quantity 5 g
Formula Weight 174
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.