missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobiphenyl, 98%, Thermo Scientific™

Product Code. 10576023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10576023 50 g Glass bottle 50g
10566023 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10576023 Supplier Thermo Scientific Acros Supplier No. 373270500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41604-19-7
Molecular Formula C12H8BrF
Molecular Weight (g/mol) 251.10
MDL Number MFCD00051716
InChI Key HTRNHWBOBYFTQF-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobiphenyl, 4-bromo-2-fluoro-1,1'-biphenyl, 2-fluoro-4-bromo biphenyl, 1,1'-biphenyl, 4-bromo-2-fluoro, 4-bromo-2-fluorodiphenyl, 4-bromo-2-fluoro-1,1-biphenyl, 4-bromo-2-fluoro biphenyl, pubchem2652, acmc-209jkv, 2-fluoro-4-bromobiphenyl
PubChem CID 521063
IUPAC Name 4-bromo-2-fluoro-1-phenylbenzene
SMILES FC1=CC(Br)=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H3(F)Br
Quantity 50 g
Solubility Information Solubility in water: insoluble
Formula Weight 251.1
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Bromo-2-fluorobiphenyl
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobiphenyl
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.