missing translation for 'onlineSavingsMsg'
Learn More

4-bromo-2-fluorobenzylamine, 98%

Product Code. 10178152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10178152 5 g Glass bottle 5g
10629384 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10178152 Supplier Thermo Scientific Acros Supplier No. 384440050
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112734-22-2 | C7H7BrFN | 204.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112734-22-2
Molecular Formula C7H7BrFN
Molecular Weight (g/mol) 204.04
MDL Number MFCD00153076
InChI Key RLTFBWCBGIZCDQ-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzylamine, 4-bromo-2-fluorophenyl methanamine, 2-fluoro-4-bromobenzylamine, 2-fluoro-4-bromobenzyl amine, 4-bromo-2-fluorobenzyl amine, benzenemethanamine, 4-bromo-2-fluoro, 1-4-bromo-2-fluorophenyl methanamine, 4-bromo-2-fluoro-phenyl methanamine, 4-bromo-2-fluorophenyl methylamine, pubchem4709
PubChem CID 3770848
IUPAC Name (4-bromo-2-fluorophenyl)methanamine
SMILES C1=CC(=C(C=C1Br)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NH2CH2C6H3(F)Br
Refractive Index 1.5630 to 1.5650 (20°C, 589nm)
Quantity 5 g
Formula Weight 204.04
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorobenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.