missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoyl chloride, 99%

Product Code. 10793203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793203 25 g Glass bottle 25g
10170172 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10793203 Supplier Thermo Scientific Acros Supplier No. 347460250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 151982-51-3 | C7H3BrClFO | 237.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151982-51-3
Molecular Formula C7H3BrClFO
Molecular Weight (g/mol) 237.46
MDL Number MFCD03411584
InChI Key PCFIABOQFAFDAU-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chloride
PubChem CID 2734026
IUPAC Name 4-bromo-2-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1Br)F)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 24.0°C to 24.5°C
Color White to Yellow
Boiling Point 86.0°C to 88.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: hydrdolyses
Formula Weight 237.46
Percent Purity 99%
Physical Form Crystalline Mass or Liquid
Chemical Name or Material 4-Bromo-2-fluorobenzoyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2-fluorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.