missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoyl chloride, 98%

Product Code. 11334596
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334596 25 g 25g
11314596 1 g 1g
11324596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11334596 Supplier Thermo Scientific Alfa Aesar Supplier No. L20142.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 151982-51-3 | C7H3BrClFO | 237.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151982-51-3
Molecular Formula C7H3BrClFO
Molecular Weight (g/mol) 237.452
MDL Number MFCD03411584
InChI Key PCFIABOQFAFDAU-UHFFFAOYSA-N
PubChem CID 2734026
IUPAC Name 4-bromo-2-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1Br)F)C(=O)Cl

Specifications

Melting Point 24°C to 25°C
Boiling Point 86°C to 88°C (15 mmHg)
Refractive Index 1.5877
Quantity 25 g
UN Number UN3261
Beilstein 6323925
Sensitivity Moisture sensitive
Synonym 2-fluoro-4-bromobenzoyl chloride, benzoyl chloride, 4-bromo-2-fluoro, benzoyl chloride,4-bromo-2-fluoro, acmc-20amnm, 4-bromo-2-fluorobenzoylchloride, 4-bromo-2-fluoro-benzoyl chloride, 4-bromo-2-fluorobenzoic acid chloride, 4-bromanyl-2-fluoranyl-benzoyl chloride
Formula Weight 237.45
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.