missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorobenzoic acid, 98%

Product Code. 11420641
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420641 5 g 5g
11430641 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420641 Supplier Thermo Scientific Alfa Aesar Supplier No. B20118.06
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112704-79-7 | C7H3BrFO2 | 218.00 g/mol

4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2-fluorobenzoic acid may be used in the synthesis of biaryl intermediates by palladium-mediated coupling with various aryl boronic acids

Solubility
Solubility in methanol, very faint turbidity.

Notes
Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112704-79-7
Molecular Formula C7H3BrFO2
Molecular Weight (g/mol) 218.00
MDL Number MFCD00143263
InChI Key ZQQSRVPOAHYHEL-UHFFFAOYSA-M
Synonym 2-fluoro-4-bromobenzoic acid, 4-bromo-2-fluoro-benzoic acid, 4-bromo-2-fluorobenzoicacid, benzoic acid, 4-bromo-2-fluoro, pubchem1313, enzalutamide impurity a, acmc-1btmf, 2-fluro-4-bromobenzoic acid, ksc174m4p, rarechem al bo 0754
PubChem CID 302681
IUPAC Name 4-bromo-2-fluorobenzoic acid
SMILES [O-]C(=O)C1=CC=C(Br)C=C1F

Specifications

Melting Point 210°C to 213°C
Quantity 5 g
Beilstein 6323931
Formula Weight 219.01
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.