missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoroanisole, 97%

Product Code. 11404373
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404373 5 g 5g
11414373 25 g 25g
11424373 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404373 Supplier Thermo Scientific Alfa Aesar Supplier No. A10948.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2357-52-0 | C7H6BrFO | 205.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2357-52-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD00011710
InChI Key DWNXGZBXFDNKOR-UHFFFAOYSA-N
Synonym 4-bromo-2-fluoroanisole, 2-fluoro-4-bromoanisole, 1-bromo-3-fluoro-4-methoxybenzene, benzene, 4-bromo-2-fluoro-1-methoxy, 2-fluoro-4-bromo anisol, 2-fluoro-4-bromo anisole, 4-bromo-2-fluoro-1-methoxy-benzene, 4-bromo-2-fluorophenylmethyl ether, 4-brom-2-fluor-1-methoxybenzol, pubchem1971
PubChem CID 75378
IUPAC Name 4-bromo-2-fluoro-1-methoxybenzene
SMILES COC1=C(C=C(C=C1)Br)F

Specifications

Density 1.59
Boiling Point 208°C
Flash Point 98°C (208°F)
Refractive Index 1.545
Quantity 5 g
Beilstein 3240714
Formula Weight 205.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-fluoroanisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.