missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluoro-1,1'-biphenyl, TRC

Product Code. 30337505
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337505 10 g 10g
30334387 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30337505 Supplier Toronto Research Chemicals Supplier No. TRCB68440510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-2-fluoro-1,1'-biphenyl
CAS 41604-19-7
Molecular Formula C12H8BrF
Quantity 10 g
Synonym 2-Fluoro-4-bromobiphenyl, 4-Bromo-2-fluorobiphenyl, 4'-Bromo-2'-fluorobiphenyl
InChI Formula InChI=1S/C12H8BrF/c13-10-6-7-11(12(14)8-10)9-4-2-1-3-5-9/h1-8H
SMILES Fc1cc(Br)ccc1c2ccccc2
IUPAC Name 4-bromo-2-fluoro-1-phenylbenzene
Molecular Weight (g/mol) 251.09
Formula Weight 249.9793
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.