missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-ethylaniline, 97%

Product Code. 11373336
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373336 25 g 25g
11363336 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373336 Supplier Thermo Scientific Alfa Aesar Supplier No. L12725.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 45762-41-2 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45762-41-2
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00041434
InChI Key LGOZNQPHTIGMQJ-UHFFFAOYSA-N
Synonym benzenamine, 4-bromo-2-ethyl, 2-ethyl-4-bromoaniline, acmc-20amnq, pubchem23174, 4-bromo-2-ethyl-aniline, 4-bromo-2-ethylbenzenamine, 4-bromo-2-ethylphenylamine, 4-bromanyl-2-ethyl-aniline, 4-bromo-2-ethylaniline
PubChem CID 2801283
IUPAC Name 4-bromo-2-ethylaniline
SMILES CCC1=C(C=CC(=C1)Br)N

Specifications

Density 1.412
Boiling Point 110°C to 112°C (2 mmHg)
Refractive Index 1.602
Quantity 25 g
UN Number UN2810
Beilstein 2802710
Formula Weight 200.08
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-ethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.