missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 15462079
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15462079 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15462079 Supplier Thermo Scientific Alfa Aesar Supplier No. H64753.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as an primary and secondary intermediate for pharmaceutical, organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885702-34-1
Molecular Formula C4H2BrClN2
Molecular Weight (g/mol) 193.43
MDL Number MFCD08272196
InChI Key FBEBVAQOMVWORE-UHFFFAOYSA-N
Synonym pyrimidine, 4-bromo-2-chloro-9ci, pyrimidine, 4-bromo-2-chloro, 2-chloro-4-bromopyrimidine
PubChem CID 45117394
IUPAC Name 4-bromo-2-chloropyrimidine
SMILES ClC1=NC=CC(Br)=N1

Specifications

Quantity 250 mg
Beilstein 10334333
Solubility Information Slightly soluble in water.
Formula Weight 193.43
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chloropyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.