missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chloropyridine, 94%

Product Code. 11312369
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312369 25 g 25g
11302369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312369 Supplier Thermo Scientific Alfa Aesar Supplier No. H27299.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73583-37-6 | C5H3BrClN | 192.44 g/mol

It is used in the synthesis of quaterpyridine nemertelline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of quaterpyridine nemertelline.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73583-37-6
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD03840756
InChI Key ONHMWUXYIFULDO-UHFFFAOYSA-N
Synonym 2-chloro-4-bromopyridine, pyridine, 4-bromo-2-chloro, 4-bromo-2-chloro-pyridine, pubchem1171, acmc-1bjmw, ksc381c4t, tpc-py061, 4-bromo-2-chloropyridine, abbypharma ap-18-5387, 4-bromo-2-chloropyridine 5g
PubChem CID 3847770
IUPAC Name 4-bromo-2-chloropyridine
SMILES C1=CN=C(C=C1Br)Cl

Specifications

Melting Point 18°C to 20°C
Density 1.734
Boiling Point 70°C (3 mmHg)
Flash Point 104°C (219°F)
Quantity 25 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 192.44
Percent Purity 94%
Chemical Name or Material 4-Bromo-2-chloropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.