missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorobenzonitrile, 96%

Product Code. 11342886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342886 5 g 5g
11332886 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342886 Supplier Thermo Scientific Alfa Aesar Supplier No. L10602.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 154607-01-9 | C7H3BrClN | 216.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154607-01-9
Molecular Formula C7H3BrClN
Molecular Weight (g/mol) 216.46
MDL Number MFCD00040883
InChI Key AYQBMZNSJPVADT-UHFFFAOYSA-N
Synonym 2-chloro-4-bromobenzonitrile, 4-bromo-2-chloro-benzonitrile, benzonitrile, 4-bromo-2-chloro, 4-bromo-2-chlorobenzenecarbonitrile, pubchem12801, intermediates-zcf02177, acmc-209db5, ksc497g8d
PubChem CID 7010420
IUPAC Name 4-bromo-2-chlorobenzonitrile
SMILES ClC1=C(C=CC(Br)=C1)C#N

Specifications

Melting Point 67°C to 72°C
Boiling Point 142°C to 143°C (10 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 3240885
Formula Weight 216.47
Percent Purity 96%
Chemical Name or Material 4-Bromo-2-chlorobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.