missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-2'-chloroacetanilide, 98+%

Product Code. 11402948
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402948 25 g 25g
11412948 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11402948 Supplier Thermo Scientific Alfa Aesar Supplier No. A18652.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3460-23-9 | C8H7BrClNO | 248.504 g/mol

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Soluble in hot methanol (very faint turbidity).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3460-23-9
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.504
MDL Number MFCD00040852
InChI Key MITWNEIUIPGZKR-UHFFFAOYSA-N
Synonym 4'-bromo-2'-chloroacetanilide, n-4-bromo-2-chlorophenyl acetamide, 4-bromo-2-chloroacetanilide, acetamide, n-4-bromo-2-chlorophenyl, maybridge3_007422, 2-chlor-4-bromacetanilid, acmc-209i8u, buttpark 83\07-60, acetanilide, 4'-bromo-2'-chloro
PubChem CID 610167
IUPAC Name N-(4-bromo-2-chlorophenyl)acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)Br)Cl

Specifications

Melting Point 151°C to 152°C
Quantity 25 g
Beilstein 2719133
Solubility Information Soluble in hot methanol (very faint turbidity).
Formula Weight 248.51
Percent Purity ≥98%
Chemical Name or Material 4'-Bromo-2'-chloroacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.