missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-dimethylaniline, 98%

Product Code. 11459562
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Unit Size:
10g
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459562 50 g 50g
11443484 10 g 10g
11379419 25 g 25g
11469562 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11459562 Supplier Thermo Scientific Alfa Aesar Supplier No. A10712.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24596-19-8 | C8H10BrN | 200.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24596-19-8
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00007826
InChI Key QGLAYJCJLHNIGJ-UHFFFAOYSA-N
Synonym 4-bromo-2,6-xylidine, benzenamine, 4-bromo-2,6-dimethyl, 4-bromo-2,6-dimethyl-phenylamine, 2-amino-5-bromo-m-xylene, 2,6-dimethyl-4-bromoaniline, 4-bromo-2,6-dimethylbenzenamine, 4-bromo-2,6-dimethylphenylamine, 4-bromo-2,6-dimethyl-aniline, 4-bromo-2,6-dimethylaniline, 4-bromo-2,6-dimethyl-benzenamine
PubChem CID 90549
IUPAC Name 4-bromo-2,6-dimethylaniline
SMILES CC1=CC(Br)=CC(C)=C1N

Specifications

Melting Point 47°C to 52°C
Quantity 50 g
UN Number UN2811
Beilstein 1936300
Boiling Point 120°C (0.2 mmHg)
Formula Weight 200.08
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,6-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.