missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzyl alcohol, 97%

Product Code. 15408178
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15408178 1 g 1g
15418178 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15408178 Supplier Thermo Scientific Alfa Aesar Supplier No. H61623.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 162744-59-4 | C7H5BrF2O | 223.017 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162744-59-4
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.017
MDL Number MFCD03094461
InChI Key LSRHFWSNUFIKER-UHFFFAOYSA-N
Synonym 4-bromo-2,6-difluorobenzyl alcohol, 4-bromo-2,6-difluorophenyl methanol, 4-bromo-2,6-difluorobenzylalcohol, benzenemethanol, 4-bromo-2,6-difluoro, 4-bromo-2,6-difluoro-benzenemethanol, 2,6-difluoro-4-bromobenzyl alcohol, 4-bromo-2,6-difluoro-phenyl methanol, 2,6-difluoro-4-bromo benzyl alcohol, pubchem4882, acmc-209dox
PubChem CID 2773303
IUPAC Name (4-bromo-2,6-difluorophenyl)methanol
SMILES C1=C(C=C(C(=C1F)CO)F)Br

Specifications

Melting Point 76°C to 81°C
Quantity 1 g
Formula Weight 223.01
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-2,6-difluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.