missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzotrifluoride, 98%

Product Code. 15421579
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421579 1 g 1g
15441579 250 mg 250mg
15431579 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421579 Supplier Thermo Scientific Alfa Aesar Supplier No. H64386.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 156243-64-0 | C7H2BrF5 | 260.989 g/mol

It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

Solubility
Slightly soluble in water.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156243-64-0
Molecular Formula C7H2BrF5
Molecular Weight (g/mol) 260.989
MDL Number MFCD01631616
InChI Key QPJKIRNIIXIPIE-UHFFFAOYSA-N
PubChem CID 23450919
IUPAC Name 5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1F)C(F)(F)F)F)Br

Specifications

Color Colorless
Quantity 1 g
Beilstein 13252396
Synonym 3,5-difluoro-4-trifluoromethyl bromobenzene, 5-bromo-1,3-difluoro-2-trifluoromethyl benzene, 4-bromo-2,6-difluorobenzotrifluoride, benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl, acmc-1cioh, intermediates-zcf02186, 2,6-difluoro-4-bromobenzotrifluoride, 1-bromo-3,5-difluoro-4-trifluoromethylbenzene, 3,5-difluoro-4-trifluoromethylbromobenzene, benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
Solubility Information Slightly soluble in water.
Formula Weight 260.99
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-2,6-difluorobenzotrifluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.