missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,6-difluorobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog Number p-7055736
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
156243-64-0
C7H2BrF5
260.989
MFCD01631616
QPJKIRNIIXIPIE-UHFFFAOYSA-N
23450919
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br
This item is not returnable. View return policy
156243-64-0
C7H2BrF5
260.989
MFCD01631616
QPJKIRNIIXIPIE-UHFFFAOYSA-N
23450919
5-bromo-1,3-difluoro-2-(trifluoromethyl)benzene
C1=C(C=C(C(=C1F)C(F)(F)F)F)Br

It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the lewis acid-promoted synthesis of unsymmetrical and highly functionalized carbazoles and dibenzofurans from biaryl triazenes.

Solubility
Slightly soluble in water.

Notes
Store away from bases and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat.
TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mg
Beilstein 13252396
Synonym 3,5-difluoro-4-trifluoromethyl bromobenzene, 5-bromo-1,3-difluoro-2-trifluoromethyl benzene, 4-bromo-2,6-difluorobenzotrifluoride, benzene, 5-bromo-1,3-difluoro-2-trifluoromethyl, acmc-1cioh, intermediates-zcf02186, 2,6-difluoro-4-bromobenzotrifluoride, 1-bromo-3,5-difluoro-4-trifluoromethylbenzene, 3,5-difluoro-4-trifluoromethylbromobenzene, benzene,5-bromo-1,3-difluoro-2-trifluoromethyl
Solubility Information Slightly soluble in water.
Formula Weight 260.99
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Bromo-2,6-difluorobenzotrifluoride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'