missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluorophenol, 99%

Product Code. 11346639
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11346639 5 g 5g
11356639 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346639 Supplier Thermo Scientific Alfa Aesar Supplier No. H26212.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 486424-36-6 | C6H3BrF2O | 208.99 g/mol

4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-difluorophenol is used as a raw material in organic synthesis. Used in the synthesis allyloxy-based biphenyl liquid crystals with multiple lateral fluoro substituents.

Solubility
Soluble in water (slightly).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486424-36-6
Molecular Formula C6H3BrF2O
Molecular Weight (g/mol) 208.99
MDL Number MFCD04973751
InChI Key BYZMZJIWCQTYSR-UHFFFAOYSA-N
Synonym 1-bromo-2,5-difluoro-4-hydroxybenzene, 4-bromo-2,5-difluoro-phenol, phenol, 4-bromo-2,5-difluoro, acmc-20amnw, phenol,4-bromo-2,5-difluoro, 4-bromanyl-2,5-bis fluoranyl phenol, 1-hydroxy-4-bromo-2,5-difluoro-benzene
PubChem CID 7018042
IUPAC Name 4-bromo-2,5-difluorophenol
SMILES C1=C(C(=CC(=C1F)Br)F)O

Specifications

Melting Point 51°C to 53°C
Quantity 5 g
Solubility Information Soluble in water (slightly).
Formula Weight 208.99
Percent Purity 99%
Chemical Name or Material 4-Bromo-2,5-difluorophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.