missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-difluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 10163044 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163044 1 g Glass bottle 1g
10674355 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10163044 Supplier Thermo Scientific Acros Supplier No. 399880010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 207974-14-9
Molecular Formula C6H2BrClF2O2S
Molecular Weight (g/mol) 291.49
MDL Number MFCD00042181
InChI Key SBMKFWMFNIEPDN-UHFFFAOYSA-N
Synonym 4-bromo-2,5-difluorobenzene-1-sulfonylchloride, 4-bromo-2,5-difluorobenzenesulphonyl chloride, 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride, 4-bromo-2,5-difluorophenyl chlorosulfone, 4-bromo-2,5-difluorobenzenesulphonylchloride, benzenesulfonyl chloride, 4-bromo-2,5-difluoro, 4-bromo-3,6-difluoro-benzenesulfonylchloride, pubchem5058, acmc-209feg, 4-bromo-2,5-difluorobenzenesulfonylchloride
PubChem CID 574590
IUPAC Name 4-bromo-2,5-difluorobenzenesulfonyl chloride
SMILES FC1=CC(=C(F)C=C1Br)S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 39°C
Color Orange or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula F2C6H2(Br)SO2Cl
Quantity 1 g
Formula Weight 291.49
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromo-2, 5-difluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-2,5-difluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.