missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(4-morpholinyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 15448488
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260762-06-8
Molecular Formula C11H11BrN2O
Molecular Weight (g/mol) 267.126
InChI Key SEDCSLWVDKMJCF-UHFFFAOYSA-N
Synonym 4-bromo-2-morpholin-4-yl benzonitrile, 4-bromo-2-morpholinobenzonitrile, acmc-209b1e
PubChem CID 70699567
IUPAC Name 4-bromo-2-morpholin-4-ylbenzonitrile
SMILES C1COCCN1C2=C(C=CC(=C2)Br)C#N

Specifications

Quantity 250 mg
UN Number UN3439
Formula Weight 267.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-2-(4-morpholinyl)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'