missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene, 99%, ACROS Organics™

Product Code. 10615711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10615711 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10615711 Supplier Thermo Scientific Acros Supplier No. 293780050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17823-46-0
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7BrF7
Refractive Index 1.4275 to 1.4295 (20°C, 589nm)
MDL Number MFCD00075240
Quantity 5 g
Synonym 1-bromo-2,3,5,6-tetrafluoro-4-trifluoromethyl benzene, 4-bromo-2,3,5,6-tetrafluorobenzotrifluoride, p-bromoheptafluorotoluene, 4-trifluoromethyl-2,3,5,6-tetrafluorobromobenzene, acmc-209ee7, 4-bromoheptafluorotoluene, 4-bromo-alpha,alpha,alpha,2,3,5,6-heptafluorotoluene, 1-trifluoromethyl-2,3,5,6-tetrafluoro-4-bromobenzene
InChI Key DONVNRFILRLHJB-UHFFFAOYSA-N
SMILES C1(=C(C(=C(C(=C1F)F)Br)F)F)C(F)(F)F
IUPAC Name 1-bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol) 296.97
PubChem CID 626637
Formula Weight 296.96
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 4-Bromo-α,α,α,2,3,5,6-heptafluorotoluene, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.