missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole, 98+%

Product Code. p-7055953
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
Unit Size:
1kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421979 25 g 25g
15431979 500 g 500g
15441979 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421979 Supplier Thermo Scientific Alfa Aesar Supplier No. H64673.14
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2075-45-8 | C3H3BrN2 | 146.98 g/mol

4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4?-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4′-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2075-45-8
Molecular Formula C3H3BrN2
Molecular Weight (g/mol) 146.98
MDL Number MFCD00075602
InChI Key WVGCPEDBFHEHEZ-UHFFFAOYSA-N
Synonym 4-bromopyrazole, 1h-pyrazole, 4-bromo, 4-brompyrazole, 4-bromo-pyrazole, pyrazole, 4-bromo, 4-bromo pyrazole, byz, zlchem 126, 1h-pyrazole,4-bromo, pubchem11041
PubChem CID 16375
IUPAC Name 4-bromo-1H-pyrazole
SMILES BrC1=CNN=C1

Specifications

Melting Point 93°C to 96°C (literature)
Boiling Point 250°C to 260°C (literature)
Quantity 25 g
Beilstein 106510
Solubility Information Slightly soluble in water.
Formula Weight 146.97
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 4-Bromo-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.