missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole-5-carbaldehyde, 95+%, Thermo Scientific™

Product Code. 10442654
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10442654 5 g 5g
10550353 250 mg 250mg
10344202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10442654 Supplier Thermo Scientific Maybridge Supplier No. RF03950DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 287917-97-9
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.985
InChI Key UWGFONONBAIDAF-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-5-carboxaldehyde, 4-bromo-2h-pyrazole-3-carbaldehyde, 4-bromo-1h-pyrazole-3-carboxaldehyde, 1h-pyrazole-3-carboxaldehyde,4-bromo, 1h-pyrazole-3-carboxaldehyde, 4-bromo, 4-bromopyrazole-5-carbaldehyde, 4-bromo-5-formyl-1h-pyrazole, 4-bromo-pyrazole-3-carboxaldehyde, 4-bromo-2h-pyrazole-3-carboxaldehyde
PubChem CID 2782154
IUPAC Name 4-bromo-1H-pyrazole-5-carbaldehyde
SMILES C1=NNC(=C1Br)C=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 174.98
Percent Purity 95%
Chemical Name or Material 4-Bromo-1H-pyrazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.