missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-pyrazole-3-carbonitrile, 97%, Thermo Scientific™

Product Code. p-2701499
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10273612

Brand: Thermo Scientific Maybridge RF05036CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 288246-16-2
Molecular Formula C4H2BrN3
Molecular Weight (g/mol) 171.985
MDL Number MFCD00105683
InChI Key ZXVFKQRZKKGVNJ-UHFFFAOYSA-N
Synonym 4-bromo-1h-pyrazole-3-carbonitrile, 4-bromo-2h-pyrazole-3-carbonitrile, 4-bromopyrazole-3-carbonitrile, 4-bromo-3-cyano-1h-pyrazole, 1h-pyrazole-3-carbonitrile, 4-bromo, pubchem10182, acmc-20aa8h, maybridge1_006470, ksc497k2j, buttpark 181\04-37
PubChem CID 2735623
IUPAC Name 4-bromo-1H-pyrazole-5-carbonitrile
SMILES C1=NNC(=C1Br)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 171.98
Percent Purity 97%
Chemical Name or Material 4-Bromo-1H-pyrazole-3-carbonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.