missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1H-indole-3-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10136992
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10136992 1 g 1g
10142804 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10136992 Supplier Thermo Scientific Maybridge Supplier No. MO07344DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98600-34-1
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
InChI Key IPAFHZDRYAWZOB-UHFFFAOYSA-N
Synonym 4-bromoindole-3-carboxaldehyde, 4-bromoindole-3-carboxyaldehyde, 4-bromo-3-formylindole, 4-bromo-1h-indole-3-carboxaldehyde, 4-bromoindole-3-carbaldehyde, 4-bromo-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 4-bromo, 4-bromo-3-indolecarbaldehyde, pubchem7682, acmc-209saq
PubChem CID 2763178
IUPAC Name 4-bromo-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C(=C1)Br)C(=CN2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 224.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-1H-indole-3-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.