missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Product Code. 10420564
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10420564 5 g Amber Glass Bottle 5g
10136772 250 mg Amber Glass Bottle 250mg
10433773 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10420564 Supplier Thermo Scientific Maybridge Supplier No. MO00978DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 497832-99-2
Molecular Formula C5H4BrF3N2
Molecular Weight (g/mol) 229
MDL Number MFCD04115415
InChI Key TZAYNGPUOOUEAP-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-3-trifluoromethyl-1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl pyrazole, 1h-pyrazole,4-bromo-1-methyl-3-trifluoromethyl, 1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl, pubchem16983, buttpark 99\18-60, 4-bromo-1-methyl-3-trifluoromethyl pyrazol, 4-bromo-1-methyl-3-trifluoromethyl ;-1h-pyrazole
PubChem CID 2794651
IUPAC Name 4-bromo-1-methyl-3-(trifluoromethyl)pyrazole
SMILES CN1C=C(C(=N1)C(F)(F)F)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 229
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.