missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde, 97%

Product Code. 15426547
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15426547 1 g 1g
15428595 250 mg 250mg
15436547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15426547 Supplier Thermo Scientific Alfa Aesar Supplier No. H33690.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 473528-88-0 | C5H5BrN2O | 189.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 473528-88-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD03074341
InChI Key GXAHYXQWHWDEDY-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazole-5-carbaldehyde, 4-bromo-1-methylpyrazole-5-carbaldehyde, 4-bromo-2-methyl-2h-pyrazole-3-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 4-bromo-1-methyl, acmc-1akfs, 4-bromo-1-methylpyrazole-5-carboxaldehyde, 4-bromo-1-methyl-1h-pyrazole-5-carboxaldehyde, 4-bromo-2-methyl-2 h-pyrazole-3-carbaldehyde
PubChem CID 2794634
IUPAC Name 4-bromo-2-methylpyrazole-3-carbaldehyde
SMILES CN1C(=C(C=N1)Br)C=O

Specifications

Melting Point 97°C to 98°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-5-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.