missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10091784
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10091784 5 g 5g
10039431 250 mg 250mg
10212552 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091784 Supplier Thermo Scientific Maybridge Supplier No. MO00866DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 473528-88-0
Molecular Formula C5H5BrN2O
Molecular Weight (g/mol) 189.012
MDL Number MFCD03074341
InChI Key GXAHYXQWHWDEDY-UHFFFAOYSA-N
PubChem CID 2794634
IUPAC Name 4-bromo-2-methylpyrazole-3-carbaldehyde
SMILES CN1C(=C(C=N1)Br)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Synonym 4-bromo-1-methyl-1h-pyrazole-5-carbaldehyde, 4-bromo-1-methylpyrazole-5-carbaldehyde, 4-bromo-2-methyl-2h-pyrazole-3-carbaldehyde, 1h-pyrazole-5-carboxaldehyde, 4-bromo-1-methyl, acmc-1akfs, 4-bromo-1-methylpyrazole-5-carboxaldehyde, 4-bromo-1-methyl-1h-pyrazole-5-carboxaldehyde, 4-bromo-2-methyl-2 h-pyrazole-3-carbaldehyde
Formula Weight 189.01
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-1H-pyrazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.