missing translation for 'onlineSavingsMsg'
Learn More

(4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol, ≥97%, Thermo Scientific™

Product Code. 10265404
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10265404 5 g 5g
10694765 250 mg 250mg
10265884 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265404 Supplier Thermo Scientific Maybridge Supplier No. CC63909DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-65-2
Molecular Formula C5H7BrN2O
Molecular Weight (g/mol) 191.028
MDL Number MFCD06202888
InChI Key JFGLJTFTVBZOCB-UHFFFAOYSA-N
Synonym 4-bromo-1-methyl-1h-pyrazol-3-yl methanol, 4-bromo-1-methylpyrazol-3-yl methanol, 4-bromo-3-hydroxymethyl-1-methyl-1h-pyrazole, 4-bromo-1-methylpyrazol-3-yl methan-1-ol, pubchem23717
PubChem CID 24229713
IUPAC Name (4-bromo-1-methylpyrazol-3-yl)methanol
SMILES CN1C=C(C(=N1)CO)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Quantity 5 g
Formula Weight 191.03
Percent Purity 97%
Chemical Name or Material (4-Bromo-1-methyl-1H-pyrazol-3-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.