missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butyne, 97%

Product Code. 15460108
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15460108

Brand: Thermo Scientific Alfa Aesar H53367.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38771-21-0 | C4H5Br | 132.988 g/mol

4-Bromo-1-butyne is used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-1-butyne is used for pharmaceutical intermediates.

Solubility
Immiscible with water.

Notes
Recommended storage in refrigerator. Keep away from sources of ignition. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38771-21-0
Molecular Formula C4H5Br
Molecular Weight (g/mol) 132.988
MDL Number MFCD10000883
InChI Key XLYOGWXIKVUXCL-UHFFFAOYSA-N
Synonym 4-bromo-1-butyne, 1-bromo-3-butyne, 1-butyne, 4-bromo, 4-bromobutyne, 3-butynyl bromide, 1-bromo-3-butynol, 4-bromo-but-1-yne, acmc-1afup, 1-butyne, 4-bromo-6ci,9ci
PubChem CID 11073464
IUPAC Name 4-bromobut-1-yne
SMILES C#CCCBr

Specifications

Density 1.417
Boiling Point 110°C
Flash Point 24°C (75°F)
Linear Formula HC≡CCH2CH2Br
Refractive Index 1.481
Quantity 25 g
UN Number UN1992
Solubility Information Immiscible with water.
Formula Weight 132.99
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-butyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.