missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butyl Acetate, TRC

Product Code. 30348418
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348418 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348418 Supplier Toronto Research Chemicals Supplier No. TRCB681965100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromo-1-butyl Acetate
CAS 4753-59-7
Molecular Formula C6H11BrO2
Quantity 100 g
Synonym 4-Bromo-1-butanol 1-Acetate, 4-Bromo-1-butanol Acetate, 1-Acetoxy-4-bromobutane, 4-Acetoxy-1-bromobutane, 4-Acetoxybutyl Bromide, 4-Bromo-1-acetoxybutane, 4-Bromo-1-butanol Acetate, 4-Bromobutyl Acetate, Acetic Acid 4-Bromobutyl Ester, NSC 33919
InChI Formula InChI=1S/C6H11BrO2/c1-6(8)9-5-3-2-4-7/h2-5H2,1H3
SMILES CC(=O)OCCCCBr
IUPAC Name 4-bromobutyl acetate
Molecular Weight (g/mol) 195.05
Formula Weight 193.9942
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.