missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-1-butene, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-1-butene |
| CAS | 5162-44-7 |
| Molecular Formula | C4H7Br |
| Quantity | 1 g |
| Synonym | 1-Bromo-3-butene, 3-Butenyl bromide, 4-Bromobutene-1, Allylcarbinyl Bromide, Homoallyl Bromide |
| InChI Formula | InChI=1S/C4H7Br/c1-2-3-4-5/h2H,1,3-4H2 |
| SMILES | BrCCC=C |
| IUPAC Name | 4-bromobut-1-ene |
| Molecular Weight (g/mol) | 135 |
| Formula Weight | 133.9731 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction