missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-butanol, 85+%

Product Code. 10571514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571514 1 g Glass bottle 1g
10637264 5 g Glass bottle 5g
10465465 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571514 Supplier Thermo Scientific Acros Supplier No. 390010010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 33036-62-3 | C4H9BrO | 153.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33036-62-3
Molecular Formula C4H9BrO
Molecular Weight (g/mol) 153.02
MDL Number MFCD00015387
InChI Key SIJLYRDVTMMSIP-UHFFFAOYSA-N
Synonym 4-bromo-1-butanol, 1-butanol, 4-bromo, 4-bromobutanol, 4-bromo-butan-1-ol, 1-bromo-4-hydroxybutane, tetramethylene bromohydrin, 4-bromo-n-butanol, 1-bromobutan-4-ol, 4-bromo butan-1-ol, acmc-1axcv
PubChem CID 118388
IUPAC Name 4-bromobutan-1-ol
SMILES C(CCBr)CO

Specifications

CAS Min % 85.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.0680g/mL
Boiling Point 56.0°C to 58.0°C (2.0 mmHg)
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 85% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)4OH
Quantity 1 g
Specific Gravity 1.068
Solubility Information Solubility in water: insoluble
Formula Weight 153.02
Percent Purity 85+%
Physical Form Liquid
Chemical Name or Material 4-Bromo-1-butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Bromo-1-butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.