missing translation for 'onlineSavingsMsg'
Learn More

4-(BOC-amino)-1-butanol, 98%

Product Code. 10047849 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass Bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10047849 5 mL Glass Bottle 5mL
10109739 1 mL Glass Bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10047849 Supplier Thermo Scientific Acros Supplier No. 434210050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75178-87-9 | C9H19NO3 | 189.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75178-87-9
Molecular Formula C9H19NO3
Molecular Weight (g/mol) 189.26
MDL Number MFCD01861334
InChI Key LIYMTLVBAVHPBU-UHFFFAOYSA-N
Synonym tert-butyl 4-hydroxybutyl carbamate, tert-butyl n-4-hydroxybutyl carbamate, 4-boc-amino-1-butanol, 4-tert-butoxycarbonylamino-1-butanol, carbamic acid, 4-hydroxybutyl-, 1,1-dimethylethyl ester, tert-butyl 4-hydroxybutylcarbamate, n-boc-4-aminobutanol, butylhydroxybutylcarbamate, acmc-1bfe1, 4-t-boc-amino-1-butanol
PubChem CID 545141
IUPAC Name tert-butyl N-(4-hydroxybutyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCCO

Specifications

Melting Point 37°C to 40°C
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Density 1.0200g/mL
Quantity 5 mL
Formula Weight 189.25
Percent Purity 98%
Specific Gravity 1.02
Physical Form Low Melting Solid or Liquid
Chemical Name or Material 4-(Boc-Amino)-1-butanol
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Boc-amino)-1-butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.