missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Boc-2-oxopiperazine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 4-Boc-2-oxopiperazine |
| CAS | 76003-29-7 |
| Molecular Formula | C9H16N2O3 |
| Quantity | 1 g |
| Synonym | 1,1-Dimethylethyl 3-oxo-1-piperazinecarboxylate, 1-(tert-Butoxycarbonyl)piperazin-3-one, 1-Boc-3-oxo-piperazine, 1-tert-Butoxycarbonyl-3-oxopiperazine, 2-Oxo-4-(tert-butoxycarbonyl)piperazine, 3-Oxo-1-piperazinecarboxylic Acid 1,1-Dimethylethyl Ester, 3-Oxopiperazine-1-carboxylic Acid tert-Butyl Ester, 3-Oxopiperazine-1-carboxylic Acid tert-Butyl Ester, 4-(tert-Butyloxycarbonyl)-2-piperazinone, 4-tert-Butoxycarbonylpiperazin-2-one, tert-Butyl 3-oxo-1-piperazinecarboxylate |
| InChI Formula | InChI=1S/C9H16N2O3/c1-9(2,3)14-8(13)11-5-4-10-7(12)6-11/h4-6H2,1-3H3,(H,10,12) |
| SMILES | CC(C)(C)OC(=O)N1CCNC(=O)C1 |
| IUPAC Name | tert-butyl 3-oxopiperazine-1-carboxylate |
| Molecular Weight (g/mol) | 200.23 |
| Formula Weight | 200.1161 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction