Learn More
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%
Description
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifikationer
Specifikationer
| Melting Point | 83°C to 87°C |
| Color | White to Yellow |
| Quantity | 25 g |
| UN Number | UN2811 |
| Solubility Information | Soluble in water. |
| Formula Weight | 342.23 |
| Percent Purity | 97% |
| Physical Form | Powder |
| Chemical Name or Material | 4-Boc-1-(5-bromo-2-pyridyl)piperazine |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.