missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine, TRC

Product Code. 30347318
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30347318 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347318 Supplier Toronto Research Chemicals Supplier No. TRCB62510010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine
CAS 1076199-22-8
Molecular Formula C18 H24 N2 O5
Quantity 10 mg
InChI Formula InChI=1S/C18H24N2O5/c1-18(2,3)25-17(22)20-10-8-19(9-11-20)16(21)15-12-23-13-6-4-5-7-14(13)24-15/h4-7,15H,8-12H2,1-3H3
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C(=O)C2COc3ccccc3O2
IUPAC Name tert-butyl 4-(2,3-dihydro-1,4-benzodioxine-3-carbonyl)piperazine-1-carboxylate
Molecular Weight (g/mol) 348.39
Formula Weight 348.1685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.