missing translation for 'onlineSavingsMsg'
Learn More

4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8, TRC

Product Code. 30335694
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335694 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335694 Supplier Toronto Research Chemicals Supplier No. TRCB6251025MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Boc-1-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8
CAS 1189968-99-7
Molecular Formula C18H16D8N2O5
Quantity 5 mg
Synonym N-Boc-N’-(1,4-benzodioxan-2-ylcarbonyl)piperazine-d8, 4-[(2,3-Dihydro-1,4-benzodioxin-2-yl)carbonyl]-1-piperazinecarboxylic Acid 1,1-Dimethylethyl Ester-d8
InChI Formula InChI=1S/C18H24N2O5/c1-18(2,3)25-17(22)20-10-8-19(9-11-20)16(21)15-12-23-13-6-4-5-7-14(13)24-15/h4-7,15H,8-12H2,1-3H3/i8D2,9D2,10D2,11D2
SMILES [2H]C1([2H])N(C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])N(C(=O)C2COc3ccccc3O2)C1([2H])[2H]
IUPAC Name tert-butyl 2,2,3,3,5,5,6,6-octadeuterio-4-(2,3-dihydro-1,4-benzodioxine-3-carbonyl)piperazine-1-carboxylate
Molecular Weight (g/mol) 356.44
Formula Weight 356.2187
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.