missing translation for 'onlineSavingsMsg'
Learn More

4-(Bis(4-Hydroxyphenyl)methylene)cyclohexa-2,5-dienone, TRC

Product Code. 30138629
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138629 500 mg 500mg
30136560 1 g 1g
30003276 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138629 Supplier Toronto Research Chemicals Supplier No. TRCB434150500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19 H14 O3
Molecular Weight (g/mol) 290.31
Synonym Aurin, 4,4'-Dihydroxyfuchsone, Rosalic Acid, Pararosolic Acid, Spirit Aurine, Corallin, Aurin 555, C.I. 43800, 4,4'-Dihydroxyfuchsone, Corallin Spirit Soluble;
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES Oc1ccc(cc1)C(=C2C=CC(=O)C=C2)c3ccc(O)cc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H14O3/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,20-21H
Formula Weight 290.0943
Chemical Name or Material 4-(Bis(4-Hydroxyphenyl)methylene)cyclohexa-2,5-dienone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.